Learn More
CAPRICO BIOTECHNOLOGIES INC CD22 APC/IFluor 700 MyG13 ASR
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4051175
Encompass_Preferred
CD22 APC iFluor 700 MyG13 ASR is a monoclonal antibody conjugated to APC iFluor 700 dye, produced by Invitrogen. It is clone MyG13 and specifically binds to human CD22. This antibody is recommended for flow cytometry analysis of human CD22 expression on cells. It has an excitation max of 695nm and emission max of 719nm. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100μg of antibody.
Catalog No. 50-227-9067