Learn More
CAPRICO BIOTECHNOLOGIES INC CD22 PE Antibody, 4051025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4051025
Encompass_Preferred
The Caprico CD22 PE Antibody is a mouse monoclonal antibody that binds to human CD22. It is conjugated to R-phycoerythrin (PE) and is intended for use in flow cytometry. This antibody recognizes an extracellular epitope and does not cross-react with other cell surface antigens. It can be used to identify and characterize B lineage cells in bone marrow, blood, and lymphoid tissues. The antibody is supplied as a liquid in PBS with 0.1% sodium azide and 2 mg/ml BSA. It has a concentration of 0.2 mg/ml and is available in sizes of 100 tests and 1000 tests.
Catalog No. 50-227-9066