Learn More
Biolegend Purified anti-human CD223 (LAG-3)
Supplier:  Biolegend 369202
Encompass_Preferred
Purified anti-human CD223 (LAG-3) [7H2C65]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. NC1448483