Learn More
Aviva Systems Biology Corp CD226 Antibody - C-terminal region (ARP63749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD226. It was validated on Western Blot. . Alternate names: DNAM-1, DNAM1, PTA1, TLiSA1ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rat
Catalog No. 50-288-072