Learn More
Sino Biological CD226 / DNAM-1 Antibody (APC), Rabbit MAb 25Tests
Supplier:  Sino Biological 50232R026A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD226 / DNAM-1 (rM CD226 / DNAM-1; Catalog#50232-M08H; NP_848802.2; Met1-Pro254) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-222-2211