Learn More
Genscript Corporation CD229/SLAMF3Biotin His Av
Supplier:  Genscript Corporation Z04424500
Encompass
CD229 was strongly and homogeneously overexpressed on the PC of patients with monoclonal gammopathy of undetermined significance (MGUS) smoldering myeloma MM and PC leukemia CD229 was particularly overexpressed on those PC showing an abnormal phenotype such as expression of CD56 Most importantly CD229 was also highly expressed on those cells in the patients BM displaying the phenotype of chemotherapy-resistant and myeloma-propagating cells
Catalog No. 50-000-58986