Learn More
ACROBiosystems Recombinant Protein;Human CD229 / SLAMF3 Protein;HEK293;100UG;CD9,CD229 antigen,CD229,cell-surface molecule Ly-9,hly9,Ly9,lymphocyte antigen 9Cell surface molecule Ly-9,Mly9,SLAMF3,T-lymphocyte surface antigen Ly-9
Supplier:  ACROBiosystems CD9H52H6100UG
Encompass_Preferred
Recombinant Protein;Human CD229 / SLAMF3 Protein;100UG;Human CD229 (CD9-H52H6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7567