Learn More
CAPRICO BIOTECHNOLOGIES INC CD23 APC/IFluor700
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4095175
Encompass_Preferred
The CD23 APC-iFluor 700 antibody from Caprico Biotechnologies is a monoclonal antibody that targets human CD23. It is conjugated to APC-iFluor 700 and is suitable for flow cytometry applications. This antibody recognizes the low affinity IgE receptor CD23, also known as Fc epsilon RII. It can be used to identify B cells, follicular dendritic cells, and other CD23 expressing cells by flow cytometry. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.
Catalog No. 50-227-9071