Learn More
Medchemexpress LLC CD23/Fc epsilon RII 100ug
SDP
Supplier:  Medchemexpress LLC HYP74321100UG
Encompass_Preferred
CD23/Fc epsilon RII Protein a low-affinity receptor for IgE and CR2/CD21 regulates IgE production and B cell differentiation On B cells it initiates IgE-dependent antigen uptake and presentation to T cells On macrophages IgE binding induces intracellular killing of parasites through the L-Arginine-nitric oxide pathway CD23/Fc epsilon RII homotrimer interacts with IGHE to regulate IgE homeostasis and with CR2/CD21 for binding CD23/Fc epsilon RII Protein Mouse (HEK293 His) is the recombinant mouse-derived CD23/Fc epsilon RII protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-78806