Learn More
Enzo Life Sciences CD23 (human) monoclonal antibody (9p25) (Alexa Fluor 647) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2460100
Encompass_Preferred
Clone: 9p25. Isotype: IgG1. Immunogen: EBV transformed lymphoblastoid cell line. Species reactivity: Human. Specificity: Recognizes human B lymphocytes. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2792