Learn More
Enzo Life Sciences CD23 (human) monoclonal antibody (9p25) (biotin conjugate) (100µg)
Supplier: Enzo Life Sciences ENZABS2470100
Clone: 9p25. Isotype: IgG1. Immunogen: EBV transformed lymphoblastoid cell line. Species reactivity: Human. Specificity: Recognizes human B lymphocytes. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.