Learn More
CAPRICO BIOTECHNOLOGIES INC CD235a Antibody, Clone HI264, APC
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4043045
Encompass_Preferred
The CD235a Antibody Clone HI264 APC is an antibody that binds to human CD235a, also known as glycophorin A. It is conjugated to APC fluorochrome and is suitable for flow cytometry applications. This monoclonal antibody recognizes a sialoglycoprotein expressed on erythrocytes. It can be used for research purposes to identify erythroid lineage cells by flow cytometry. The antibody is available in 100 tests per vial.
Catalog No. 50-227-9077