Learn More
ACROBiosystems Recombinant Protein;Human CD23 / Fc epsilon RII Protein;HEK293;1MG;CD3,FCER2,FCER-2,CD23,CD-23,CD23A,CD-23A,CLEC4J,FCE2,FCE-2,IGEBF,FcεRII,FcεRⅡ,Fcr,Fcgr,Fc receptor,Fc epsilon receptor
Catalog No. 501657491
Encompass_Preferred
Recombinant Protein;Human CD23 / Fc epsilon RII Protein;1MG;Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accession # P06734-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 32.9 kDa. The protein migrates as 40-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7491 Supplier ACROBiosystems Supplier No. CD3H52491MG
Recombinant Protein;Human CD23 / Fc epsilon RII Protein;1MG;Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accession # P06734-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 32.9 kDa. The protein migrates as 40-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.