Learn More
CAPRICO BIOTECHNOLOGIES INC CD24 Antibody, 4233045, APC Conjugated
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4233045
Encompass_Preferred
The CD24 Antibody (4233045) APC-conjugated from Caprico Biotechnologies is a monoclonal antibody that specifically binds to human CD24. It is conjugated to Allophycocyanin (APC) and is recommended for detection of CD24 protein in flow cytometry. This antibody is produced in mouse and has been tested in flow cytometry applications. The immunogen is a recombinant human CD24 protein. According to the manufacturer, this antibody has high affinity and specificity for human CD24. The APC fluorochrome has an excitation max of 633-635 nm and emission max of 660-680 nm. This product is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Recommended usage is 5 ul per test.
Catalog No. 50-227-9083