Learn More
CAPRICO BIOTECHNOLOGIES INC CD24 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4233095
Encompass_Preferred
The CD24 antibody is a mouse monoclonal antibody that recognizes human CD24. It is unconjugated and has been tested for flow cytometry, immunohistochemistry, and ELISA applications. The antibody reacts with the small heavily glycosylated mucin-like cell surface protein CD24 expressed on granulocytes, B cells, and many cancer cells. This antibody is purified by protein A/G and is supplied at 0.5 mg/ml in PBS with 0.09% sodium azide.
Catalog No. 50-227-9081