Learn More
ACROBiosystems ACROBiosystems Human CD24 Protein, Fc Tag
Catalog No. 501956585
Encompass_Preferred
Recombinant Protein;100UG;Human CD24, Fc Tag (CD4-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD24, Fc Tag (Cat. No. CD4-H5254) at 10 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 2-78 ng/mL (QC tested).
Catalog No. 50-195-6585 Supplier ACROBiosystems Supplier No. CD4H5254100UG
Recombinant Protein;100UG;Human CD24, Fc Tag (CD4-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD24, Fc Tag (Cat. No. CD4-H5254) at 10 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 2-78 ng/mL (QC tested).