Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD24 Protein, Fc,Avitag™
Catalog No. 501956563
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD24, Fc,Avitag (CD4-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 31.4 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD24, Fc,Avitag (Cat. No. CD4-H82F5) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-CD24 MAb (SN3) with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-195-6563 Supplier ACROBiosystems Supplier No. CD4H82F5200UG
Recombinant Protein;200UG;Biotinylated Human CD24, Fc,Avitag (CD4-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 31.4 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD24, Fc,Avitag (Cat. No. CD4-H82F5) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-CD24 MAb (SN3) with a linear range of 0.6-20 ng/mL (QC tested).