Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD24 Protein, His,Avitag™
Catalog No. 501956550
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 6.7 kDa. The protein migrates as 18-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD24, His,Avitag (Cat. No. CD4-H82E9) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-CD24 MAb (SN3) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-195-6550 Supplier ACROBiosystems Supplier No. CD4H82E925UG
Add to Cart
Add to Cart
Recombinant Protein;25UG;Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 6.7 kDa. The protein migrates as 18-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD24, His,Avitag (Cat. No. CD4-H82E9) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-CD24 MAb (SN3) with a linear range of 0.6-10 ng/mL (QC tested).