Learn More
ACROBiosystems ACROBiosystems Human CD24 Protein, Mouse IgG2a Fc Tag
Catalog No. 501956555
Encompass_Preferred
Recombinant Protein;100UG;Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 30.0 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD24, Mouse IgG2a Fc Tag (Cat. No. CD4-H5257) at 5 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 20-313 ng/mL (QC tested).
Catalog No. 50-195-6555 Supplier ACROBiosystems Supplier No. CD4H5257100UG
Recombinant Protein;100UG;Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 30.0 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD24, Mouse IgG2a Fc Tag (Cat. No. CD4-H5257) at 5 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 20-313 ng/mL (QC tested).