Learn More
Sino Biological Human CD24 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 11030H05H100
Encompass_Preferred
A DNA sequence encoding the human CD24 (NP_037362.1) (Met1-Gly59) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-211-4760