Learn More
Medchemexpress LLC CD24 Rat HEK293 F 10ug
SDP
Catalog No. 5000178456
Encompass_Preferred
The CD24 protein plays a critical role in cellular differentiation in various cell types possibly triggered by binding of lectin-like ligands to the CD24 carbohydrate This signal is transduced through second messengers released from GPI anchors CD24 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD24 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-78456 Supplier Medchemexpress LLC Supplier No. HYP7431810UG
The CD24 protein plays a critical role in cellular differentiation in various cell types possibly triggered by binding of lectin-like ligands to the CD24 carbohydrate This signal is transduced through second messengers released from GPI anchors CD24 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD24 protein expressed by HEK293 with C-hFc labeled tag