Learn More
Aviva Systems Biology Corp CD244 Antibody - C-terminal region (ARP61376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD244. It was validated on Western Blot. . Alternate names: 2B4, NAIL, NKR2B4, Nmrk, SLAMF4ApplicationWB. Species reactivity: Human
Catalog No. 50-287-356