Learn More
CAPRICO BIOTECHNOLOGIES INC CD25 Antibody, 4026-045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4026045
Encompass_Preferred
The Caprico CD25 Antibody is a rabbit monoclonal antibody that recognizes human CD25. It is recommended for detection of CD25 protein in Western blot, immunohistochemistry, and flow cytometry applications. The immunogen is a recombinant protein corresponding to amino acids 20-250 of human CD25. This antibody has been tested in human, mouse, and rat samples. According to the product details, it detects a band of 55 kDa in Western blots. The concentration is 0.5 mg/mL, with 100 μg in 200 μL. Overall, this monoclonal antibody can be used to detect expression of the CD25 protein across various applications.
Catalog No. 50-227-9088