Learn More
CAPRICO BIOTECHNOLOGIES INC CD25 PE, 4026025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4026025
Encompass_Preferred
The CD25 PE Antibody from Caprico Biotechnologies is a phycoerythrin-conjugated mouse monoclonal antibody that recognizes human CD25. CD25, also known as the interleukin-2 receptor alpha chain or IL-2Rα, is a 55 kDa transmembrane protein expressed on activated T and B lymphocytes, regulatory T cells, and some thymocytes. This antibody is recommended for detection of CD25 expression on human lymphocytes by flow cytometry. It has been tested on peripheral blood lymphocytes. Each vial contains 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The recommended dilution is 1:10. This antibody is a useful reagent for studying T cell activation and regulatory T cells.
Catalog No. 50-227-9087