Learn More
Sino Biological Recombinant Human CD25/IL-2R alpha Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10165H27HB100
Encompass_Preferred
A DNA sequence encoding the human IL2RA (NP_000408.1) (Met1-Cys213) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5235