Learn More
Cayman Chemical CD25InTRleukIn2Ra ChImRIc
SDP
Catalog No. 502919454
Encompass_Preferred
Immunogen: Recombinant human CD25/IL-2Rα • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human, cynomolgus monkey, rhesus monkey • Cross Reactivity: (+) CD25 • Applications: FC, IF, IHC, WB
Catalog No. 50-291-9454 Supplier Cayman Chemical Supplier No. 37170200 UG
Immunogen: Recombinant human CD25/IL-2Rα • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human, cynomolgus monkey, rhesus monkey • Cross Reactivity: (+) CD25 • Applications: FC, IF, IHC, WB