Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 939MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: LPFS2/1611. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Human peripheral blood lymphocytes. Human tonsil and lymph node. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CD22 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 120kDa.Chromosome: 12p13. Gene Symbol: CD27. Gene ID: 939. Ensemble: ENSG00000139193. Uniprot: XP_016875723.
Catalog No. 50-188-5338