Learn More
Biolegend Brilliant Violet 510™ anti-human CD27
Supplier:  Biolegend 302835
Encompass_Preferred
Brilliant Violet 510™ anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. NC1291014