Learn More
Aviva Systems Biology Corp CD27 Antibody - C-terminal region (ARP59093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD27. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CD27. Alternate names: MGC20393, S152, T14, TNFRSF7, Tp55ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-575