Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD27 / TNFRSF7 Protein, Fc,Avitag™
Catalog No. 501957377
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD27, Fc,Avitag (TN7-H82F6) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # NP_001233).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.7 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) with a linear range of 5-156 ng/mL (QC tested).;Flow Cytometry assay shows that Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) can bind to Raji cells Surface Human CD70. The concentration of CD27 used is 10 μg/mL (Routinely tested).
Catalog No. 50-195-7377 Supplier ACROBiosystems Supplier No. TN7H82F6200UG
Recombinant Protein;200UG;Biotinylated Human CD27, Fc,Avitag (TN7-H82F6) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # NP_001233).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.7 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) with a linear range of 5-156 ng/mL (QC tested).;Flow Cytometry assay shows that Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) can bind to Raji cells Surface Human CD70. The concentration of CD27 used is 10 μg/mL (Routinely tested).