Learn More
Genscript Corporation CD27 Ligand/CD70 His Cynomol
Supplier:  Genscript Corporation Z04112100
Encompass
CD70 also named CD27 ligand (CD27L) is a type II transmembrane glycoprotein belonging to the TNF superfamily (TNFSF) and has been designated TNFSF7 CD70 is a cytokine that binds to CD27 Plays a role in T-cell activation Induces the proliferation of costimulated T-cells and enhances the generation of cytolytic T-cells
Catalog No. 50-000-57777