Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems CD7C52591MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Arg 191 (Accession # XP_005569963.1). In the region Ala 20 - Arg 191, the AA sequence of Cynomolgus and Rhesus macaque CD27 are homologus.;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-C5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-7233