Learn More
Enzo Life Sciences CD27 Recombinant monoclonal antibody (RM27-3E5) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8000200
Encompass_Preferred
Clone: RM27-3E5. Isotype: IgGκ. Immunogen: This antibody was raised by immunising mouse with mouse CD27-human IgG1 Fc fusion protein. Isolated popliteal lymph node cells were then fused with P3U1 myeloma cells to produce stable hybridomas. Source: This chimeric rabbit antibody was made using the variable domain sequences of the original Rat IgG2a format, for improved compatibility with existing reagents, assays and techniques. Species reactivity: Mouse. Applications: ELISA. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3324