Learn More
Genscript Corporation CD27/TNFRSF7 mFc Chimera Mous
Catalog No. 5000059304
Encompass
CD27 also known as TNFRSF7 is an approximately 55 kDa transmembrane protein in the TNF receptor superfamily It functions as a co stimulatory molecule that supports lymphocyte activation and survival It binds to ligand CD70 and plays a key role in regulating B-cell activation and immunoglobulin synthesis
Catalog No. 50-000-59304 Supplier Genscript Corporation Supplier No. Z04308100
CD27 also known as TNFRSF7 is an approximately 55 kDa transmembrane protein in the TNF receptor superfamily It functions as a co stimulatory molecule that supports lymphocyte activation and survival It binds to ligand CD70 and plays a key role in regulating B-cell activation and immunoglobulin synthesis