Learn More
AdipoGen CD274 [B7-H1/PD-L1] (human):Fc (human) (rec.) (non-lytic)
SDP
Catalog No. 501147422
Encompass_Preferred
CD274 [B7-H1/PD-L1] (human):Fc (human) (rec.) (non-lytic), AdipoGen Life Sciences. Protein. The ed of human CD274 [B7-H1] (aa 19-238) is fused to the N-terminus of the Fc region of a mutant human IgG1. Shows the biological function of the CD274 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain. Lyophilized from 0.2?m-filtered solution in PBS.
Catalog No. 50-114-7422 Supplier AdipoGen Supplier No. CHIHF220PDL1C100
CD274 [B7-H1/PD-L1] (human):Fc (human) (rec.) (non-lytic), AdipoGen Life Sciences. Protein. The ed of human CD274 [B7-H1] (aa 19-238) is fused to the N-terminus of the Fc region of a mutant human IgG1. Shows the biological function of the CD274 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain. Lyophilized from 0.2?m-filtered solution in PBS.