Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD276/B7-H3 4Ig /B7 1mg
SDP

Catalog No. 5000185028
Encompass_Preferred

Recombinant human CD276/B7-H3 (4Ig) protein is expressed in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized material. It is glycosylated and provided at high purity for use in immune regulation, tumor biology, and assay development where mammalian post-translational modification is required.

  • Mammalian-expressed, glycosylated protein suitable for functional and binding assays.
  • C-terminal His tag enables detection and affinity purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Multiple vial sizes available from 5 μg to 1 mg to fit assay scale.
  • Lyophilized formulation for improved stability and handling.
  • Store at -20°C; aliquot and keep at -20°C or -80°C for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O to maintain activity.

Catalog No. 50-001-85028 Supplier Medchemexpress LLC Supplier No. HYP706521MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human CD276/B7-H3 (4Ig) protein is expressed in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized material. It is glycosylated and provided at high purity for use in immune regulation, tumor biology, and assay development where mammalian post-translational modification is required.

  • Mammalian-expressed, glycosylated protein suitable for functional and binding assays.
  • C-terminal His tag enables detection and affinity purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Multiple vial sizes available from 5 μg to 1 mg to fit assay scale.
  • Lyophilized formulation for improved stability and handling.
  • Store at -20°C; aliquot and keep at -20°C or -80°C for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O to maintain activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.