Learn More
AdipoGen CD279 [PD-1] (human) (rec.) (His),Protein
SDP
Supplier:  AdipoGen CHIHF201PD1C100
Encompass_Preferred
CD279 [PD-1] (human) (rec.) (His), AdipoGen Life Sciences. Protein. The extracellular domain of human CD279 [PD-1] (aa 25-167) is fused at the C-terminus to a His-tag. Measured by its binding ability in a functional ELISA. Lyophilized from 0.2¿m-filtered solution in PBS.
Catalog No. 50-117-6271