Learn More
ACROBiosystems Recombinant Protein;Human CD27 / TNFRSF7 Protein;HEK293;100UG;CD7,CD27,TNFRSF7,S152,T14,Tp55
Supplier:  ACROBiosystems CD7H522B100UG
Encompass_Preferred
Recombinant Protein;Human CD27 / TNFRSF7 Protein;100UG;Human CD27 (CD7-H522b) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # P26842-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-165-7493