Learn More
ACROBiosystems ACROBiosystems Human CD27 Ligand / CD70 Protein, Fc Tag (active trimer) (MALS verified)
Catalog No. 501956442
Encompass_Preferred
Recombinant Protein;100UG;Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 74.1 kDa. The protein migrates as 130-150 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27, His Tag (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5266) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-6442 Supplier ACROBiosystems Supplier No. CDLH5266100UG
Recombinant Protein;100UG;Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5266) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 74.1 kDa. The protein migrates as 130-150 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27, His Tag (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5266) with a linear range of 0.2-8 ng/mL (QC tested).