Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD27 Ligand / CD70 Protein, His,Avitag™,Flag Tag (active trimer) (MALS verified)
Supplier:  ACROBiosystems CDLH82D7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD27 Ligand, His,Avitag,Flag Tag (active trimer) (MALS verified) (CDL-H82D7) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™) and a flag tag. The protein has a calculated MW of 51.0 kDa. The protein migrates as 47-57 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27, Fc Tag (Cat. No. CD7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, His,Avitag,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H82D7) with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-195-6422