Learn More
ACROBiosystems Recombinant Protein;Mouse CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CDL,CD70,CD27LG,TNFSF7,TNFSF7G,CD27L,CD27-L,CD27-LG
Catalog No. 501657988
Encompass_Preferred
Recombinant Protein;Mouse CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 47 - Pro 195 (Accession # O55237-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 (Cat. No. CD7-H522b) at 1 μg/mL (100 μL/well) can bind Mouse CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-M5251) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-165-7988 Supplier ACROBiosystems Supplier No. CDLM5251100UG
Recombinant Protein;Mouse CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 47 - Pro 195 (Accession # O55237-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 (Cat. No. CD7-H522b) at 1 μg/mL (100 μL/well) can bind Mouse CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-M5251) with a linear range of 0.3-2 ng/mL (QC tested).