Learn More
ACROBiosystems Recombinant Protein;Human CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;CDL,CD70,CD27LG,TNFSF7,TNFSF7G,CD27L,CD27-L,CD27-LG
Supplier:  ACROBiosystems CDLH525A1MG
Encompass_Preferred
Recombinant Protein;Human CD27 Ligand / CD70 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H525a) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Pro 193 (Accession # P32970-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 44.0 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Human CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525a) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-7970