Learn More
ACROBiosystems Recombinant Protein;Mouse CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CD7,CD27,TNFRSF7,S152,T14,Tp55
Supplier:  ACROBiosystems CD7M5252100UG
Encompass_Preferred
Recombinant Protein;Mouse CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-M5252) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Arg 182 (Accession # P41272-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-M5252) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse CD70, His Tag, Avi Tag with a linear range of 1-16 ng/mL (QC tested).;Immobilized Mouse CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-M5252) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 1-16 ng/mL (Routinely tested).
Catalog No. 50-165-8041