Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CD7,CD27,TNFRSF7,S152,T14,Tp55
Catalog No. 501658088
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Arg 191 (Accession # XP_005569963.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-C5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-165-8088 Supplier ACROBiosystems Supplier No. CD7C5259100UG
Recombinant Protein;Cynomolgus / Rhesus macaque CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Arg 191 (Accession # XP_005569963.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-C5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2-31 ng/mL (QC tested).