Learn More
ACROBiosystems Recombinant Protein;Human CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CD7,CD27,TNFRSF7,S152,T14,Tp55
Catalog No. 501658032
Encompass_Preferred
Recombinant Protein;Human CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # P26842-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 1 μg/mL (100 μL/well) can bind Human CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-H5257) with a linear range of 0.005-0.078 μg/mL (QC tested).
Catalog No. 50-165-8032 Supplier ACROBiosystems Supplier No. CD7H5257100UG
Recombinant Protein;Human CD27 / TNFRSF7 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # P26842-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.2 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 1 μg/mL (100 μL/well) can bind Human CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-H5257) with a linear range of 0.005-0.078 μg/mL (QC tested).