Learn More
ACROBiosystems CD28 1mg
Supplier:  ACROBiosystems CD8H52H31MG
Encompass_Preferred
Human / Cynomolgus / Rhesus macaque CD28 Protein His Tag (CD8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # P10747-1).
Catalog No. 50-253-5336