Learn More
ACROBiosystems ACROBiosystems Biotinylated Human / Cynomolgus / Rhesus macaque CD28 Protein, Fc,Avitag™
Catalog No. 501957444
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhesus macaque CD28 are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (Cat. No. CD8-H82F2) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 5-78 ng/mL (QC tested).;
Catalog No. 50-195-7444 Supplier ACROBiosystems Supplier No. CD8H82F2200UG
Recombinant Protein;200UG;Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhesus macaque CD28 are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (Cat. No. CD8-H82F2) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 5-78 ng/mL (QC tested).;