Learn More
Medchemexpress LLC CD28 Mouse HEK293 50ug
SDP
Catalog No. 5000185106
Encompass_Preferred
Recombinant mouse CD28 extracellular fragment produced in mammalian cells for immunology research. The protein comprises residues N20-K149 and includes C-terminal human Fc and 6xHis tags to facilitate purification and detection. It is intended for use in studies of T-cell costimulation, ligand binding, and signaling, and is supplied with low endotoxin and manufacturer handling and storage recommendations.
Catalog No. 50-001-85106 Supplier Medchemexpress LLC Supplier No. HYP7061450UG
Recombinant mouse CD28 extracellular fragment produced in mammalian cells for immunology research. The protein comprises residues N20-K149 and includes C-terminal human Fc and 6xHis tags to facilitate purification and detection. It is intended for use in studies of T-cell costimulation, ligand binding, and signaling, and is supplied with low endotoxin and manufacturer handling and storage recommendations.