Learn More
Sino Biological CD28 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 17052H08H1
Encompass_Preferred
A DNA sequence encoding the Human / Cynomolgus / Rhesus CD28 (P10747-1) (Met1-Pro152) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-237-4447