Learn More
Sino Biological Recombinant Human, Cynomolgus, Rhesus CD28 Protein (ECD, Dimer, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11524H49HB20
Encompass_Preferred
A DNA sequence encoding the Human/Cynomolgus/Rhesus CD28 (P10747-1) (Met1-Pro152) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-0679